Orders LinkContactUsLinkComponent
Hitachi SEM Mount Box, Electron Microscopy Sciences
Hitachi SEM Mount Box, Electron Microscopy Sciences
EMS-Hitachi SEM Mount Box
Catalog # 102091-314
Supplier #:  76504
Hitachi SEM Mount Box, Electron Microscopy Sciences
Catalog # 102091-314
Hitachi SEM Mount Box, Electron Microscopy Sciences
Hitachi SEM Mount Box, Electron Microscopy Sciences
EMS-Hitachi SEM Mount Box
Price
$209.66
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS introduces a unique box  for Hitachi SEM mounts with M4 threaded hole in the base
The box is numbered 1 – 10 to accommodate Hitachi mounts from 15 to 32 mm diameter.
Specifications
  • Description:
    EMS-Hitachi SEM Mount Box