Micro Tissue Grinder Kit, Electron Microscopy Sciences
Product Details & Documents
This complete selection of Wheaton Micro Tissue Grinders is conveniently packed in a lightweight protective carrying case. The 0.5 ml micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures.
- Complete kit includes 7 micro tissue grinders
- Made from Wheaton 33 low extractable borosilicate glass
- Replacement parts available separately
- Autoclavable
This particular tissue grinder mortar can be safely joined with another threaded glass component if needed, using the Wheaton Connection®. The Kit is packed in high-density polyethylene case with foam inserts.
Specifications
- Description:Complete Kit