Protector® Evidence Drying Cabinet, Labconco
Product Details & Documents
The durable, stainless steel-lined Protector® Evidence Drying Cabinet provides a secure ventilated area to dry evidence such as clothing and bedding
Protector Evidence Drying Cabinet with Washdown features a spray nozzle that allows the interior to be washed down for easy clean up and after the cabinet has been properly disinfected. A built-in water pump aids clean up. Note that Evidence Drying Cabinet units with Washdown do not include UV lights.
Protector® Benchtop Evidence Drying Cabinets provide a compact, space-saving method of securely drying evidence. The cabinet's HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The key-activated ultraviolet light provides secondary disinfection of the interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing.
A variety of accessories for Protector Evidence Drying Cabinets are available, including filters, cleaning supplies, drain kits for washdown units and more. Choose from a variety of accessories for all sizes of Protector Evidence Drying Cabinets. In applicable instances, note the size of the cabinet when ordering prefilters.
The cabinet's HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The key-activated ultraviolet light provides secondary disinfection of the interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing. The lockable cabinet maintains the chain of custody. Full-length cabinets are available in 3', 4' and 5' widths and benchtop cabinets in 3' width. 3' cabinets have a single door and include 2" diameter locking casters. 4' and 5' cabinets have two doors and include fixed leveling feet.
Certifications: ETL
Specifications
- Description:Protector Evidence Drying Cabinet
- Size:5'
- Electrical:230 V (Schuko)
Accessories
Product Information
- Catalog #AvailabilityPrice
- Hanging Clips
Product Information
- Catalog #AvailabilityPrice
- Exhaust Connection Kit
Product Information
- Catalog #AvailabilityPrice
- Paper liners, tube of 25
Product Information
- Catalog #AvailabilityPrice
- Drying Cabinet Starter Kit
Product Information
- Catalog #AvailabilityPrice
- Prefilters, Inlet, Replacement
Product Information
- Catalog #AvailabilityPrice
- Paper Liners, Tube of 100
Product Information
- Catalog #AvailabilityPrice
- Security Tags
Product Information
- Catalog #AvailabilityPrice
- Drain Kit for Washdown Units
Product Information
- Catalog #AvailabilityPrice
- Prefilters, Exhaust, Replacement
Product Information
- Catalog #AvailabilityPrice
- Cabinet Prefilter, pkg. 10
Product Information
- Catalog #AvailabilityPrice
- Paper Liners for 5' EDC, pkg. 100
Product Information
- Catalog #AvailabilityPrice
- Door Prefilter, pkg. 10
Product Information
- Catalog #AvailabilityPrice
- Floor Paper Liners for 4' EDC, pkg. 100
Product Information
- Catalog #AvailabilityPrice
- Floor Paper Liners for 5' EDC, pkg. 25
Product Information
- Catalog #AvailabilityPrice
- Floor Paper Liners for 4' EDC, pkg. 25
Product Information
- Catalog #AvailabilityPrice
- Starter Kit for 5' EDC
Product Information
- Catalog #AvailabilityPrice
- Starter Kit for 4' EDC
Product Information
- Catalog #AvailabilityPrice
- Door Prefilter for 5' EDC, pkg. 6
Product Information
- Catalog #AvailabilityPrice
- Door Prefilter for 4' EDC, pkg. 6
Product Information
- Catalog #AvailabilityPrice
- Door Evidence Prefilter for 5' EDC, pkg. 10
Product Information
- Catalog #AvailabilityPrice
- Door Evidence Prefilter for 4' EDC, pkg. 10