$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CONVEX LENS DIAMETER 50MM
000000000002407478
:
Wards
CONVEX LENS DIAMETER 50MM
000000000002407478
:
Wards
:
PH0534L