VWR® Basic SPP Shoe Covers
Product 76462-940 is similar to 89233-796 but manufacturer and specifications may vary.
Product 414004-650 is similar to 89233-796 but manufacturer and specifications may vary.
Product 414004-653 is similar to 89233-796 but manufacturer and specifications may vary.
Product Details & Documents
VWR® Basic Protection SPP Shoe Covers are manufactured from strong, lightweight, and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in non-hazardous environments. These shoe covers are ideal for less critical areas or pre-gowning entry rooms.
Breathable and low lint SPP material maintains cleanroom integrity while allowing airflow
- Elastic top and bottom ensures a secure fit over various shoe sizes
- Latex-free construction is safe for users with latex sensitivities
- Ideal for non-hazardous environments
- Available with non-skid sole to minimize risk of slips in cleanroom or lab settings
- Cost-effective disposable garments eliminate laundering and reduce contamination risk
- Lot-controlled and manufactured in ISO certified facilities
VWR® Basic Protection SPP Shoe Covers are engineered to provide dependable protection in cleanrooms, laboratories, and other controlled environments where contamination control is critical. Constructed from 35 gsm spunbonded polypropylene (SPP), these covers offer a lightweight yet durable barrier against dry particulates and non-hazardous materials. The breathable, low-lint fabric ensures comfort during extended wear while maintaining the integrity of sensitive environments.
For added safety, shoe covers are available with a non-skid rubber-coated sole that enhances traction and reduces slip hazards on smooth surfaces. Elastic openings at both the top and bottom provide a secure fit over a wide range of footwear sizes, ensuring convenience and reliability. Manufactured in ISO13485:2016-certified facilities and classified as FDA Class I devices, these covers meet stringent quality and compliance standards. They are also latex-free, making them suitable for users with latex sensitivities.
Available in blue or white for easy identification, VWR® Basic Protection SPP Shoe Covers deliver a cost-effective, disposable solution for maintaining cleanliness and safety without compromising comfort or performance.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Packaging: Shoe covers are packaged 300 per case.
Specifications
- Ø:—
- Shoe size:XL
- Color:Blue
- Description:Heavy weight anti-skid shoe covers
- Height:—
- Length:17"
- Cat. No.:89233-796
- Bacterial Filtration Efficiency:—
- Cleanroom Class:10000/ISO 7
- Tearing Strength:—
- Garment application:Cleanroom|Industrial|Laboratory|Medical|General Purpose
- Gender:Unisex
- Fabric:Spunbound polypropylene
- Particle shedding (Helmke Drum):Level II
- Weight:—
- H₂O Resistance:—



