RIPA buffer, Thermo Scientific
250 mL
Supplier #: J63306-AK
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02100484
Documents
- Size:250 mL
- Cat. No.:AAJ63306-AK
- Physical Form:Liquid
- MDL No.:MFCD02100484
- Appearance:Colorless
- Storage Temperature:2°C to 8°C



