Alkaline running buffer solution, Thermo Scientific
500 mL
Supplier: Thermo Scientific Chemicals
: J63170-AP
Alkaline running buffer solution, Thermo Scientific
Catalog # AAJ63170-AP
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Size:500 mL
- Cat. No.:AAJ63170-AP
- Physical Form:Liquid
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



