Alkaline running buffer solution, Thermo Scientific
250 mL
Alkaline running buffer solution, Thermo Scientific
Catalog # AAJ63170-AK
CAS Number:
Jump to:
Discontinued
Product Details & Documents
Documents
Specifications
Specifications
- Size:250 mL
- Cat. No.:AAJ63170-AK
- Physical Form:Liquid
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



