Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
GTE buffer

GTE buffer

250 mL
Catalog # AAJ62597-AK
Supplier #:  J62597-AK
GTE buffer
Catalog # AAJ62597-AK
CAS Number:  
GTE buffer

250 mL

CAS Number:  
Price
$82.42
Log in for availability
Each (250ml)
Minimum order quantity is 1
Product Details & Documents

Storage temperature: 2 to 8°C

  • Suitable for miniprep and DNA purification

Contains 25 mM Tris, 50 mM glucose, and 10 mM EDTA.

Documents

Specifications

Specifications

  • Size:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.