PIPES, 1.0 M Buffer Solutions, Thermo Scientific
PIPES buffer solution, 1.0 M, 250 mL, Glass bottle, 7.5
Supplier: Thermo Scientific Chemicals
Supplier #: J62494-AK
PIPES, 1.0 M Buffer Solutions, Thermo Scientific
Catalog # AAJ62494-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159
Caution: Storage Temperature: 2 to 8 °C
Documents
Specifications
Specifications
- Concentration:1.0 M
- Pack type:Glass bottle
- Description:PIPES buffer solution
- Size:250 mL
- pH:7.5
- Cat. No.:AAJ62494-AK
- Physical Form:Liquid
- MDL No.:MFCD00006159
- Appearance:Colorless
- CAS No.:5625-37-6
- Storage Temperature:2°C to 8°C



