Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Danazol

500 mg
Catalog # AAJ62200-ME
Irritant
Health
 :  J62200-ME
Danazol
Catalog # AAJ62200-ME
CAS Number:  17230-88-5
Danazol

500 mg

CAS Number:  17230-88-5
Price
$268.74
Log in for availability
Each (500mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Danazol is a weak androgen. Also an anterior pituitary suppressant

Powder

Applications: Danazol is a weak androgen. Also an anterior pituitary suppressant

  • CAS Number:17230-88-5
  • Formula:C₂₂H₂₇NO₂
  • MDL Number:MFCD00056838
  • Molecular Weight:337.463 g/mol
  • Merck Index:14,2812
  • Melting Pt:218…220 °C
  • Storage Temperature:Freezer
  • EINECS:241-270-1
Specifications

Specifications

  • Size:
    500 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.