RIPA buffer-2, Thermo Scientific
250 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J62189-AK
RIPA buffer-2, Thermo Scientific
Catalog # AAJ62189-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Viscous liquid
Physical Form: Viscous liquid
Appearance: Colorless
Documents
Specifications
Specifications
- Size:250 mL
- Cat. No.:AAJ62189-AK
- Physical Form:Viscous liquid
- Appearance:Colorless
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



