Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
LDS-sample buffer (4X), non-reducing, Thermo Scientific

LDS-sample buffer (4X), non-reducing, Thermo Scientific

50 mL
Catalog # AAJ61894-AD
Supplier #:  J61894-AD
LDS-sample buffer (4X), non-reducing, Thermo Scientific
Catalog # AAJ61894-AD
CAS Number:  
LDS-sample buffer (4X), non-reducing, Thermo Scientific

50 mL

CAS Number:  
Price
$235.10
Log in for availability
Each (50ml)
Minimum order quantity is 1
Product Details & Documents

LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.

Physical Form: Liquid
Appearance: Blue

This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.

Specifications

Specifications

  • Size:
    50 mL
  • Cat. No.:
    AAJ61894-AD
  • Physical Form:
    Liquid
  • Appearance:
    Blue

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.