LDS-sample buffer (4X), non-reducing, Thermo Scientific
25 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J61894-AC
LDS-sample buffer (4X), non-reducing, Thermo Scientific
Catalog # AAJ61894-AC
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.
Physical Form: Liquid
Appearance: Blue
This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.
Specifications
Specifications
- Size:25 mL
- Cat. No.:AAJ61894-AC
- Physical Form:Liquid
- Appearance:Blue



