Sodium Citrate, 0.5 M Buffer Solution, pH 6.0, Thermo Scientific
250 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J61815-AK
Sodium Citrate, 0.5 M Buffer Solution, pH 6.0, Thermo Scientific
Catalog # AAJ61815-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00012462
Documents
Specifications
Specifications
- Size:250 mL
- Cat. No.:AAJ61815-AK
- Odor/Scent:Odorless
- Physical Form:Liquid
- MDL No.:MFCD00012462
- Appearance:Colorless
- Storage Temperature:2°C to 8°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



