Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Emodin

Emodin

100 mg
Catalog # AAJ61600-MC
Irritant
Supplier #:  J61600-MC
Emodin
Catalog # AAJ61600-MC
CAS Number:  518-82-1
Emodin

100 mg

CAS Number:  518-82-1
Price
$184.67
Log in for availability
Each (100mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A protein tyrosine kinase inhibitor

Powder

Applications: A protein tyrosine kinase inhibitor

Synonyms:  1,3,8-Trihydroxy-6-methylanthracene-9,10-dione, 1,3,8-Trihydroxy-6-methylanthraquinone
  • CAS Number:518-82-1
  • Formula:C₁₅H₁₀O₅
  • MDL Number:MFCD00001207
  • Molecular Weight:270.24 g/mol
  • Merck Index:14,3561
  • Storage Temperature:Ambient
  • EINECS:208-258-8
Specifications

Specifications

  • Size:
    100 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.