Citrate, 0.5 M Buffer Solutions, Thermo Scientific
Citrate buffer solution, 0.5 M, 500 mL, 3.0
Supplier: Thermo Scientific Chemicals
: J61391-AP
Citrate, 0.5 M Buffer Solutions, Thermo Scientific
Catalog # AAJ61391-AP
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Colourless odourless buffer solutions.
- CAS No.: 6132-04-3
- MDL No.: MFCD00150031
Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.
: Store in refrigerator. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
- Concentration:0.5 M
- Description:Citrate buffer solution
- Size:500 mL
- pH:3.0
- Cat. No.:AAJ61391-AP
- Odor/Scent:Odorless
- Physical Form:Liquid
- MDL No.:MFCD00150031
- Appearance:Colorless
- CAS No.:6132-04-3
- Storage Temperature:2°C to 8°C



