Citrate, 0.5 M Buffer Solutions, Thermo Scientific
Product Details & Documents
Colourless odourless buffer solutions.
- CAS No.: 6132-04-3
- MDL No.: MFCD00150031
Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.
Caution: Store in refrigerator. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Documents
Specifications
Specifications
- Concentration:0.5 M
- Description:Citrate buffer solution
- Size:250 mL
- pH:4.0
- Cat. No.:AAJ61249-AK
- Odor/Scent:Odorless
- Physical Form:Liquid
- MDL No.:MFCD00150031
- Appearance:Colorless
- CAS No.:6132-04-3
- Storage Temperature:2°C to 8°C
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



