Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Basic Yellow 1 ≥75%, tech.

Basic Yellow 1 ≥75%, tech.

5 g
Catalog # AAJ61043-06
Supplier #:  J61043-06
Basic Yellow 1 ≥75%, tech.
Catalog # AAJ61043-06
CAS Number:  2390-54-7
Basic Yellow 1 ≥75%, tech.

5 g

CAS Number:  2390-54-7
Price
$33.19
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

Powder

Applications: A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

Synonyms:  CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
  • CAS Number:2390-54-7
  • Formula:C₁₇H₁₉ClN₂S
  • MDL Number:MFCD00011944
  • Molecular Weight:318.86 g/mol
  • Storage Temperature:Ambient
  • EINECS:219-228-9
Specifications

Specifications

  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.