PIPES, 1.0 M Buffer Solutions, Thermo Scientific
PIPES buffer solution, 1.0 M, 500 mL, Glass bottle, 8.0
Supplier: Thermo Scientific Chemicals
: J60618-AP
PIPES, 1.0 M Buffer Solutions, Thermo Scientific
Catalog # AAJ60618-AP
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159
: Storage Temperature: 2 to 8 °C
Specifications
Specifications
- Concentration:1.0 M
- Pack type:Glass bottle
- Description:PIPES buffer solution
- Size:500 mL
- pH:8.0
- Cat. No.:AAJ60618-AP
- Physical Form:Liquid
- MDL No.:MFCD00006159
- Appearance:Colorless
- CAS No.:5625-37-6
- Storage Temperature:2°C to 8°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



