Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Phosphate, 0.2 M buffer soln., pH 4.4, Thermo Scientific

500 mL
  AAJ60467-AP
 :  J60467-AP
Phosphate, 0.2 M buffer soln., pH 4.4, Thermo Scientific
  AAJ60467-AP
 :  
Phosphate, 0.2 M buffer soln., pH 4.4, Thermo Scientific

500 mL

 :  
 
$101.92
   
Each (500ml)
 
 

Liquid

CAS No.: 7558-80-7
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00003527

 

 

  • Size:
    500 mL
  • Cat. No.:
    AAJ60467-AP
  • Physical Form:
    Liquid
  • MDL No.:
    MFCD00003527
  • CAS No.:
    7558-80-7
  • Storage Temperature:
    2°C to 8°C