Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

TBE sample buffer (6X), Thermo Scientific

50 mL
Catalog # AAJ60357-AD
 :  J60357-AD
TBE sample buffer (6X), Thermo Scientific
Catalog # AAJ60357-AD
CAS Number:  
TBE sample buffer (6X), Thermo Scientific

50 mL

CAS Number:  
Price
$86.95
Log in for availability
Each (50ml)
 
Product Details & Documents

Liquid

Physical Form: Liquid
Appearance: Blue

Specifications

Specifications

  • Size:
    50 mL
  • Cat. No.:
    AAJ60357-AD
  • Physical Form:
    Liquid
  • Appearance:
    Blue

Specifications