TBE urea sample buffer (2X), Thermo Scientific
10 mL
Supplier: Thermo Scientific Chemicals
Supplier #: J60186-AB
TBE urea sample buffer (2X), Thermo Scientific
Catalog # AAJ60186-AB
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Blue
Documents
Specifications
Specifications
- Size:10 mL
- Cat. No.:AAJ60186-AB
- Physical Form:Liquid
- Appearance:Blue



