Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

3-(4-Bromophenyl)propionic acid ≥97% for synthesis

5 g
Catalog # 101788-068
Marketsource
Supplier:   Matrix Scientific
 :  004941-5G
3-(4-Bromophenyl)propionic acid ≥97% for synthesis
Catalog # 101788-068
Supplier:  Matrix Scientific
Marketsource
CAS Number:  1643-30-7
3-(4-Bromophenyl)propionic acid ≥97% for synthesis
3-(4-Bromophenyl)propionic acid ≥97% for synthesis

5 g

CAS Number:  1643-30-7
Price
$21.54
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Hazard code(s): Irritant

  • CAS Number:1643-30-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310793
  • Molecular Weight:229.07
  • Melting Pt:133…136 °C
Specifications

Specifications

  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.