Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

RIPA buffer (2X), Thermo Scientific

125ml
  AAJ63324-EQE
 :  J63324-EQE
RIPA buffer (2X), Thermo Scientific
  AAJ63324-EQE
 :  
RIPA buffer (2X), Thermo Scientific

125ml

 :  
 
$157.08
   
Each (125ml)
 
 

Liquid

Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD02100484

 

 

  • Size:
    125ml
  • Cat. No.:
    AAJ63324-EQE
  • Physical Form:
    Liquid
  • MDL No.:
    MFCD02100484
  • Appearance:
    Colorless
  • Storage Temperature:
    2°C to 8°C