Orders LinkContactUsLinkComponent
Human/Mouse Reference RNA, Agilent Technologies
Human/Mouse Reference RNA, Agilent Technologies
Quantitative PCR Mouse Reference Total RNA, 25 µg, Pool of 11 Mouse Cell Lines Providing Broad Gene Coverage Qualified For Use in QPCR
Catalog # ST750600
Supplier #:  750600
Human/Mouse Reference RNA, Agilent Technologies
Catalog # ST750600
CAS Number:  
Human/Mouse Reference RNA, Agilent Technologies
Human/Mouse Reference RNA, Agilent Technologies
Quantitative PCR Mouse Reference Total RNA, 25 µg, Pool of 11 Mouse Cell Lines Providing Broad Gene Coverage Qualified For Use in QPCR
CAS Number:  
Price
$230.54
Log in for availability
Each
Product Details & Documents
Total RNA which has been extracted from cell lines representing different human tissues, pooled (10 human) or pooled (11 mouse), and treated to remove DNA to undetectable levels.
  • Greater consistency than RNA references derived from tissue sources
  • Maximum representation of low, medium, and high abundant gene transcripts
  • Manufactured at industrial scale lot sizes and stringently quality controlled for minimized lot-to-lot variation
  • No detectable levels of genomic DNA
Typical uses include optimization of primer/probe sets to ensure no cross reactivity, as a standard to relatively quantitate unknowns to an unchanging reference for immediate validation without wasting precious RNA, or to compare QPCR efficiencies across labs/platforms/experiments.
 : For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Description:
    Quantitative PCR Mouse Reference Total RNA
  • Size:
    25 µg
  • Details:
    Pool of 11 Mouse Cell Lines Providing Broad Gene Coverage Qualified For Use in QPCR