Human/Mouse Reference RNA, Agilent Technologies
Quantitative PCR Human Reference Total RNA, 25 µg, Pool of 10 Human Cell Lines Providing Broad Gene Coverage Qualified For Use in QPCR
: 750500
Total RNA which has been extracted from cell lines representing different human tissues, pooled (10 human) or pooled (11 mouse), and treated to remove DNA to undetectable levels.
- Greater consistency than RNA references derived from tissue sources
- Maximum representation of low, medium, and high abundant gene transcripts
- Manufactured at industrial scale lot sizes and stringently quality controlled for minimized lot-to-lot variation
- No detectable levels of genomic DNA
Typical uses include optimization of primer/probe sets to ensure no cross reactivity, as a standard to relatively quantitate unknowns to an unchanging reference for immediate validation without wasting precious RNA, or to compare QPCR efficiencies across labs/platforms/experiments.
: For research use only. Not for use in diagnostic procedures.
- Description:Quantitative PCR Human Reference Total RNA
- Size:25 µg
- Details:Pool of 10 Human Cell Lines Providing Broad Gene Coverage Qualified For Use in QPCR