Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Affinity Protein Expression and Purification Systems, Agilent Technologies

Affinity Protein Expression and Purification Systems, Agilent Technologies

Calmodulin Affinity Resin, Calmodulin binding peptide (CBP) affinity resin., 10 ml
Catalog # ST214303
Supplier #:  214303
Affinity Protein Expression and Purification Systems, Agilent Technologies
Catalog # ST214303
CAS Number:  
Affinity Protein Expression and Purification Systems, Agilent Technologies
Affinity Protein Expression and Purification Systems, Agilent Technologies

Calmodulin Affinity Resin, Calmodulin binding peptide (CBP) affinity resin., 10 ml

CAS Number:  
Price
$407.14
Log in for availability
Each (10ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.

  • pCAL vectors derived from T7 RNA polymerase-based pET vectors
  • Novel purification tag combines small size, gentle elution and high-affinity binding
  • Easy tag removal resulting in native protein

Delivery information: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).

Caution: For research use only. Not for use in diagnostic procedures.

Documents

Specifications

Specifications

  • Description:
    Calmodulin Affinity Resin
  • Size:
    10 ml
  • Details:
    Calmodulin binding peptide (CBP) affinity resin.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.