$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
MEDIUM DECBX TST BASE HIVEG 5KG TST AA
89229-372
:
HiMedia
MEDIUM DECBX TST BASE HIVEG 5KG TST AA
89229-372
:
HiMedia
:
MV912-5KG
ghs_MV912.pdf