$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
AMMONIA AC TEST KIT
89212-580
:
Lovibond
AMMONIA AC TEST KIT
89212-580
:
Lovibond
:
147211
ghs_147211.pdf
VWR GHS Compliance Letter (US)