Orders LinkContactUsLinkComponent
Comparator System 705, Lovibond®
Comparator System 705, Lovibond®
Comparator 705
Catalog # 89212-534
Supplier:   Lovibond
Supplier #:  142000
Comparator System 705, Lovibond®
Catalog # 89212-534
Supplier:  Lovibond
Comparator System 705, Lovibond®
Comparator 705
Price
$484.69
Log in for availability
Each
Product Details & Documents
The Comparator 705 is a precise, visual instrument for rapid and accurate testing. Hand-held and completely portable, can be used for testing water, wastewater, boiler and cooling water, process fluids, swimming pools, spas and more. It is ideal for the lab, plant, field or classroom.
The comparator incorporates a magnifying prism which brings the colors of the standards and test sample side-by-side and into one image, helping the user to make an exact color match.
Lovibond® has developed a line of lighting units to provide a standardized, consistent light source that simulates Northern Daylight, so reliable test results can be obtained at all times, and under all ambient light conditions.
Ordering information: Discs must be ordered separately.
Specifications
  • Description:
    Comparator 705