Orders LinkContactUsLinkComponent
Yeast Extract
Yeast Extract
Yeast extract, 100 g
Catalog # 71003-506
Supplier:   G-Biosciences
Supplier #:  RC-116
Yeast Extract
Catalog # 71003-506
Supplier:  G-Biosciences
CAS Number:  
Yeast Extract
Yeast extract, 100 g
CAS Number:  
Price
$84.97
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An autolysate of yeast cells used in preparing microbiological culture media.
Documents
Specifications
Specifications
  • Description:
    Yeast extract
  • Size:
    100 g
  • Cat. No.:
    71003-506
  • CAS No.:
    8013-01-2