$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
FLUX REMOVER ECOLINE 10OZSPRAY
14216-037
:
ITW Chemtronics
FLUX REMOVER ECOLINE 10OZSPRAY
14216-037
:
ITW Chemtronics
:
1621-10S
ghs_1621
VWR GHS Compliance Letter (US)