Orders LinkContactUsLinkComponent
EMS Engraver, Electron Microscopy Sciences
EMS Engraver, Electron Microscopy Sciences
Engraver tool stand
Catalog # 101765-016
EMS Engraver, Electron Microscopy Sciences
Catalog # 101765-016
Supplier:  
EMS Engraver, Electron Microscopy Sciences
EMS Engraver, Electron Microscopy Sciences
Engraver tool stand
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Permanently be able to mark all of your important tools and samples.
The Engraver comes with a battery pack and is operated on two "AA" batteries. Our EMS Engraver engraves test tubes, bottles, racks, or keys as well as anything made from glass, metal, stone as well as bone. Engravings are permanent and will not disappear in the presence of chemicals and solvents. The Engraver is 17.8 cm (7") in Height. We offer a stand for the EMS Engraver that is made from polypropylene and it measures (3.5 x 3.375").
Specifications
  • Description:
    Engraver tool stand