Orders LinkContactUsLinkComponent
Green-Lite™, Electron Microscopy Sciences
Green-Lite™, Electron Microscopy Sciences
Green-Lite™ 6" Round LED, Table Edge Clamp
Catalog # 101764-972
Supplier #:  68194-10
Green-Lite™, Electron Microscopy Sciences
Catalog # 101764-972
Green-Lite™, Electron Microscopy Sciences
Green-Lite™, Electron Microscopy Sciences
Green-Lite™ 6" Round LED, Table Edge Clamp
Price
$920.56
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Equipped with next generation, surface mount LEDs, these innovative designs create the new standard for illuminated magnifiers. Your investment in new technology quickly pays for itself through energy savings. Made in the USA.
  • Highest lumen output in a lighted magnifier
  • Reduced glare, heat and energy consumption
  • Multi-angle LEDs for shadowless lighting at a full range of focal lengths and heights
  • User controlled LED groups shine desired level of light exactly where required
  • Superb dimming software via convenient, reliable membrane switch
Optical features include precision ground, optical grade glass. Largest lenses in the industry. Standard 4 diopter (2x) lens - O.C. White exclusive. Optional 5 diopter (2.25x) lens available.
Electrical low voltage (15-24v) power supplies are auto-switchable with worldwide approvals. Plug in around the world by simply switching the end power cord. Energy Star compliant (each unit uses 8 watts maximum). Auto shut off at 4 hour intervals to prevent energy waste while users are away.
Mechanical high quality die cast aluminum shades and all steel arm components for ESD safety. Patented Elite arms are slim but have the greatest holding power of any arm of this type in the world. Virtually zero maintenance.
Specifications
  • Magnification:
  • Description:
    Green-Lite™ 6" Round LED, Table Edge Clamp