Orders LinkContactUsLinkComponent
VK Sponge, Contec®
VK Sponge, Contec®
VK Sponge, Single-Ply, Non sterile, 22×11×1.5 cm (8.6×4.3×0.6")
Catalog # 68310-407
Supplier:   CONTEC, INC. MX
Supplier #:  VKSPONGE-1
VK Sponge, Contec®
Catalog # 68310-407
Supplier:  CONTEC, INC. MX
VK Sponge, Contec®
VK Sponge, Contec®
VK Sponge, Single-Ply, Non sterile, 22×11×1.5 cm (8.6×4.3×0.6")
Price
$48.30
Log in for availability
Pack of 8 Case of 96
Product Details & Documents
The VertiKlean® Sponge is a disposable cleanroom sponge that features high sorbency polyester foam laminated to a Polynit wiping material with excellent abrasion resistance.
  • High sorbency polyester foam makes sponges very sorbent
  • Textured polyester surface for more aggressive cleaning tasks
  • The sponge is laser cut to completely seal the edges
Designed for general cleaning and wiping, they are ideal for use in the laboratory. Autoclavable or supplied as a sterile, ready-to-use product.
Packaging: Non sterile sponges are packaged 8/bag, 3 bags/inner bag, 4 inner bags/case. Sterile sponges are packaged 1/bag, 15 bags/inner bag, 8 inner bags/case.
Specifications
  • Description:
    VK Sponge, Single-Ply, Non sterile
  • L×W×H:
    22×11×1.5 cm (8.6×4.3×0.6")