Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Urea

Urea

5 Kg
  PAV3175
Supplier #:  V3175
Urea
  PAV3175
 :  57-13-6
Urea
Urea

5 Kg

 :  57-13-6
 
$311.39
   
Each (5kg)
 
   
 

Urea is a protein denaturant. Urea is qualified for use as the denaturing component in polyacrylamide gels.

Form: Fine, white, free-flowing pastilles
Can be stored at 15°C to 30°C (Protect from moisture)

 :  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  •  :57-13-6
  •  :NH₂CONH₂
  •  :MFCD00008022
  •  :60.06 g/mol

Documents

 

 

  • Purity:
    ≥99.0%
  • Melting Point:
    132–135°C
  • A280 at 8M in water:
    ≤0.10
  • Chloride:
    ≤0.0005%
  • Heavy Metals:
    ≤0.001%
  • Iron:
    ≤0.001%
  • Cyanate:
    none detected

 

  • Size:
    5 Kg

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.