Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cell Lysis Solution (CLA), Promega®

Cell Lysis Solution (CLA), Promega®

Cell Lysis Solution (CLA), 315 ml
Catalog # PAA7125
Supplier:   Promega Corporation
Supplier #:  A7125
Cell Lysis Solution (CLA), Promega®
Catalog # PAA7125
CAS Number:  
Cell Lysis Solution (CLA), Promega®
Cell Lysis Solution (CLA), Promega®

Cell Lysis Solution (CLA), 315 ml

CAS Number:  
Price
$100.09
Log in for availability
Each (315ml)
Minimum order quantity is 1
Product Details & Documents

Cell Lysis Solution is a component of the Wizard MagneSil, Wizard Plus and Wizard SV 96 and SV 9600 Plasmid DNA Purification Systems and the PureYield Plasmid Midiprep System.

  • Reagent Used in Plasmid DNA Isolation Protocols
  • Cat.# A7122 is a component of the Wizard(R) Plus DNA Purification Systems
  • Cat.# A7125 is a component of the PureYield™ Plasmid Midiprep System

Temperature range upto 15 °C to 30 °C

Documents

Specifications

Specifications

  • Description:
    Cell Lysis Solution (CLA)
  • Size:
    315 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.