Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Reverse Transcriptase System, Promega

Reverse Transcriptase System, Promega

Magnesium chloride solution, 25 mM, 25 mL
Catalog # PAA3513
Supplier:   Promega Corporation
Supplier #:  A3513
Reverse Transcriptase System, Promega
Catalog # PAA3513
CAS Number:  
Reverse Transcriptase System, Promega
Reverse Transcriptase System, Promega

Magnesium chloride solution, 25 mM, 25 mL

CAS Number:  
Price
$86.44
Log in for availability
Each (25ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The Reverse Transcription System provides reagents to efficiently reverse transcribe RNA into cDNA in fifteen minutes.

  • Convenient System for Reverse Transcription Using AMV Reverse Transcriptase

The Reverse Transcription System provides reagents to efficiently reverse transcribe RNA into cDNA in 15 minutes. The cDNA prepared from each reaction using this system may be used directly in multiple PCR amplifications using Taq DNA polymerase. The AMV Reverse Transcriptase synthesizes single-stranded cDNA from total or poly(A)+ RNA. Both Oligo(dT)15 and Random Primers are included, allowing cDNA synthesis from virtually any RNA source. The system contains sufficient reagents for 100 cDNA synthesis reactions, processing 1ug of RNA per reaction. Each cDNA synthesis reaction may be divided and used in up to 20 separate PCR amplifications. A polyadenylated 1.2kb RNA transcript is provided as a control template for cDNA synthesis.

Components allow for up to 100 reactions.

Accessories information: The MgCl2 and reverse transcriptase buffer are both available separately.

Ordering information: The system comes with 1500 units of AMV reverse transcriptase (HC), 2500 units of recombinant RNasin ribonuclease inhibitor, 50 µg of Oligo (dT)15 primer, 50 µg of random primers, 5 µg 1.2 kb of kanamycin positive control RNA, 320 µL of dNTP mix, 1.4 mL of reverse transcription buffer (10X), 1.2 mL of MgCl2, and 13 mL of nuclease-free water.

Documents

Specifications

Specifications

  • Description:
    Magnesium chloride solution, 25 mM
  • Size:
    25 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.