Corning® Minimum Essential Medium (Mod.) (MEM), Corning
MEM, Corning® 500 ml MEM (Minimum Essential Medium) [+] Earles salts, L-glutamine, Liquid
Supplier: Corning
Supplier #: 10-010-CV
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Catalog # 45000-380
Supplier: Corning
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.
Specifications
Specifications
- Media Format:Liquid
- Description:MEM, Corning® 500 ml MEM (Minimum Essential Medium) [+] Earles salts, L-glutamine
- Cat. No.:45000-380
- Storage Temperature:Refrigerate



