Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning

Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning

MEM, Corning® 500 ml MEM (Minimum Essential Medium) alpha medium [+] Earles salts, ribonucleosides, deoxyribonucleosides, L-glutamine, Liquid, 500 ml
Catalog # 45000-300
Supplier:   Corning
Supplier #:  10-022-CV
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Catalog # 45000-300
Supplier:  Corning
CAS Number:  
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning

MEM, Corning® 500 ml MEM (Minimum Essential Medium) alpha medium [+] Earles salts, ribonucleosides, deoxyribonucleosides, L-glutamine, Liquid, 500 ml

CAS Number:  
Price
$214.00
Log in for availability
Pack of 6 (500ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.

Certifications: Triple 0.1 µm sterilized.

Specifications

Specifications

  • Media Format:
    Liquid
  • Description:
    MEM, Corning® 500 ml MEM (Minimum Essential Medium) alpha medium [+] Earles salts, ribonucleosides, deoxyribonucleosides, L-glutamine
  • Size:
    500 ml
  • Cat. No.:
    45000-300
  • Storage Temperature:
    2...8 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.