VWR® Membrane Nitrogen Generators
4 - 60 L/min
26000-016
VWR® Membrane Nitrogen Generators are designed to supply single or multiple LC/MS instruments with dry nitrogen at purities of 99.0–99.5%.
- Recommended and certified by all major LC/MS instrument manufacturers
These membrane generators can also be used for solvent evaporation or analytical instruments. Installation requires a minimum of 60psig of compressed air to a 63.5mm (1/4") or 127mm (1/2") inlet connection. The outlet nitrogen supply is then directed to analytical instruments. The generators require no electrical connections and the only maintenance required is to change the prefilters periodically.
: Generators are also available with purity monitors. For more information, contact your Avantor sales representative.
- Flow range:4 - 60 L/min
- Cat. No.:26000-016
- Dimensions:45.7W×41.3D×130.8 H cm (18×16¹/₄×51¹/₂")
- Weight:50 kg (153 lbs.)



