Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BAG DECONAHOL 70% 32OZ STL CS8
Catalog # 14232-260
Supplier:
Veltek Associates
SDS
Expand image
BAG DECONAHOL 70% 32OZ STL CS8
Catalog # 14232-260
Supplier:
Veltek Associates
Supplier Number:
DECWFI-BAG01
SDS
Related Information
Documentation
SDS
ghs_DECWFI-B
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation