Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mouse Recombinant IL17A

Mouse Recombinant IL17A

2 x 100 µg
Catalog # 71005-132
Supplier:   IBI Scientific
Supplier #:  RPM-123
Mouse Recombinant IL17A
Catalog # 71005-132
Supplier:  IBI Scientific
CAS Number:  
Mouse Recombinant IL17A

2 x 100 µg

CAS Number:  
Price
$2,257.98
Log in for availability
Pack of 2 (100mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

IBI recombinant proteins are expressed in a yeast system (Pichia pastoris) which promotes protein folding and post-translational modifications for greater bio-activity. IBI proteins are Endotoxin FREE and are purified without HIS-TAGS or an other TAG. Our yeast expression system has proven to produce more active proteins that more closely resemble their host form.

IBI proteins are lyophilized and can be shipped at room temperature. Proteins can be reconstituted in sterile PBS with at least 0.1% of carrier protein such as BSA or cell assay media.

The recombinant mouse protein IL-17A has a gene ID of 16171. The calculated molecular weight is 15.0kDa.

Specifications

Specifications

  • Size:
    2 x 100 µg
  • Cat. No.:
    71005-132
  • Protein/Peptide Type:
    Recombinant
  • Protein/Peptide Name:
    IL17A
  • Species:
    Mouse

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.