$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CLASS 4 100G INDIVIDUAL SATIN
AP400002-226
:
Rice Lake Weighing
CLASS 4 100G INDIVIDUAL SATIN
AP400002-226
:
Rice Lake Weighing
:
RL1L13270