Orders LinkContactUsLinkComponent
SlideFolder™ Microscope Slide Holders, Simport Scientific
295×192×11 mm (11 11/16×7 9/16×7/16”), Opaque/white
Catalog # 89039-698
Supplier:   Simport Scientific
 :  M752-20WOP
SlideFolder™ Microscope Slide Holders, Simport Scientific
Catalog # 89039-698
Supplier:  Simport Scientific
SlideFolder™ Microscope Slide Holders, Simport Scientific
295×192×11 mm (11 11/16×7 9/16×7/16”), Opaque/white
Price
$210.15
Log in for availability
Case of 10
Product Details & Documents
These two-part folders hold up to twenty standard microscope slides
  • All units are stackable
  • Not autoclavable
The high-impact polystyrene base holds slides horizontally with numbered spaces for easy identification. The transparent polystyrene door covers the slides and can be swung behind the folder for space-saving purposes.
Temperature range: –80 to 80°C (–112 to 176°F)
Specifications
  • Color:
    Opaque/white
  • Dimensions:
    295×192×11 mm (11 11/16×7 9/16×7/16”)