Orders LinkContactUsLinkComponent
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
Pink
  87002-506
 :  M502-3
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
  87002-506
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
Pink
$1,064.06
Case of 1,000
These innovative disposable plastic cassettes eliminate the need for biopsy pads
Cover and base feature over 2000 square openings, each with a diameter of 0.26mm. These mesh holes allow for maximum fluid exchange. The high-density acetal polymer is resistant to the chemical action of histological solvents. The integral one-piece lids can be opened and closed multiple times. The anterior writing area is set at a 45° angle. Compatible with most cassette printers.
 : Packaged in dispenser boxes of 150 cassettes with covers assembled, four boxes per case.
  • Color:
    Pink