UNISETTE™ Processing and Embedding Cassettes, Simport
Product Details & Documents
These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The disposable biopsy cassettes have 1mm square openings and are specially designed to hold biopsy specimens during the embedding process, as well as in a storage file.
The cassettes are totally resistant to the chemical action of histological solvents. The flow-through slots allow for maximum fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap-latch and hinge-lock design prevents early separation of base and lid and allows for one-handed operation. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. The anterior writing area is at a 45° angle.
Ordering information: Cassettes are available in 11 noncytotoxic, nonmetallic colors, and three fluorescent colors. Each case contains three dispenser boxes of 500 cassettes.
Specifications
- Color:Fluorescent Pink
- Description:Standard Cassettes