$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
REACTION BLOCK SOLID 20EA/1
83007-220
:
Heidolph NA, LLC
REACTION BLOCK SOLID 20EA/1
83007-220
:
Heidolph NA, LLC
:
036190070