Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MEDIUM KOHN TWO TUBE NO2 HIVEG 100G
Catalog # 61001-302
Supplier:
HiMedia
SDS
Expand image
MEDIUM KOHN TWO TUBE NO2 HIVEG 100G
Catalog # 61001-302
Supplier:
HiMedia
Supplier Number:
MV802-100G
SDS
Related Information
Documentation
SDS
ghs_MV802.pdf
Documentation